Dihydro-N-3-oxetanyl-2H-oxetan-3-amine ≥95%
Supplier: Matrix Scientific
115879-1G
115879-10G
102899-816EA
1787.69
USD
102899-816
102899-820
Dihydro-N-3-oxetanyl-2H-oxetan-3-amine ≥95%
Dihydro-N-3-oxetanyl-2H-oxetan-3-amine
Hazard: Irritant
Formula:
C₆H₁₁NO₂ |
CAS Number:
1057682-66-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...