3,5-Difluoro-4-(oxetan-3-yloxy)benzonitrile ≥95%
Supplier: Matrix Scientific
115878-500MG
115878-1G
102898-760EA
1238.46
USD
102898-760
102898-762
3,5-Difluoro-4-(oxetan-3-yloxy)benzonitrile ≥95%
3,5-Difluoro-4-(oxetan-3-yloxy)benzonitrile
Hazard: Irritant
Formula:
C₁₀H₇F₂NO₂ |
MDL Number:
MFCD20233138 CAS Number: 1349718-43-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...