N-Cyclopropyl-3-oxetanamine ≥90%

Supplier: Matrix Scientific

115877-250MG 115877-500MG 115877-1G
102898-754EA 2172.31 USD
102898-754 102898-756 102898-758
N-Cyclopropyl-3-oxetanamine ≥90%
N-Cyclopropyl-3-oxetanamine

Hazard: Irritant

Formula: C₆H₁₁NO
MDL Number: MFCD08544351
CAS Number: 1340518-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR