3-(Chloromethyl)-3-methyloxetane ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Chloromethyl)-2-methyl-1,3-epoxypropane

115870-5G 115870-25G 115870-100G
102898-610EA 140 USD
102898-610 102898-612 102898-614
3-(Chloromethyl)-3-methyloxetane ≥95%
3-(Chloromethyl)-3-methyloxetane

Hazard: Flammable

Formula: C₅H₉ClO
MW: 120.58 g/mol
Boiling Pt: 88°C
Density: 1.1
MDL Number: MFCD01709552
CAS Number: 822-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR