3-(3-Chloro-2-fluorophenyl)-3-oxetanamine hydrochloride ≥95%
Supplier: Matrix Scientific
115868-250MG
115868-500MG
115868-1G
102898-598EA
963.08
USD
102898-598
102898-600
102898-602
3-(3-Chloro-2-fluorophenyl)-3-oxetanamine hydrochloride ≥95%
3-(3-Chloro-2-fluorophenyl)-3-oxetanamine hydrochloride
Hazard: Irritant
Formula:
C₉H₁₀Cl₂FNO |
MDL Number:
MFCD22628731 CAS Number: 1386462-30-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...