tert-Butyl-1-(oxetan-3-yl)piperidin-4-yl carbamate ≥95%

Supplier: Matrix Scientific

115867-500MG 115867-1G 115867-5G
102898-592EA 1238.46 USD
102898-592 102898-594 102898-596
tert-Butyl-1-(oxetan-3-yl)piperidin-4-yl carbamate ≥95%
tert-Butyl-1-(oxetan-3-yl)piperidin-4-yl carbamate

Hazard: Irritant

Formula: C₁₃H₂₄N₂O₃
MDL Number: MFCD20233218
CAS Number: 1228948-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR