(S)-tert-Butyl-3-(oxetan-3-ylamino)piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

115866-1G 115866-250MG
102898-590EA 2375.38 USD
102898-590 102899-804
(S)-tert-Butyl-3-(oxetan-3-ylamino)piperidine-1-carboxylate ≥95%
(S)-tert-Butyl-3-(oxetan-3-ylamino)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₃H₂₄N₂O₃
MDL Number: MFCD20233220
CAS Number: 1349702-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR