tert-Butyl-4-(oxetan-3-ylamino)piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

115864-500MG 115864-5G
102899-792EA 1512.31 USD
102899-792 102899-796
tert-Butyl-4-(oxetan-3-ylamino)piperidine-1-carboxylate ≥95%
tert-Butyl-4-(oxetan-3-ylamino)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₃H₂₄N₂O₃
MW: 256.35 g/mol
MDL Number: MFCD19982705
CAS Number: 1349718-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR