tert-Butyl-N-[(3-aminooxetan-3-yl)methyl]carbamate ≥95%

Supplier: Matrix Scientific

115862-250MG 115862-1G
102899-780EA 1044.62 USD
102899-780 102899-784
tert-Butyl-N-[(3-aminooxetan-3-yl)methyl]carbamate ≥95%
tert-Butyl-N-[(3-aminooxetan-3-yl)methyl]carbamate

Hazard: Irritant

Formula: C₉H₁₈N₂O₃
MW: 202.26 g/mol
MDL Number: MFCD19687829
CAS Number: 1379322-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR