3-(3-Bromophenyl)oxetane-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

115859-250MG 115859-500MG
102898-586EA 2267.69 USD
102898-586 102898-588
3-(3-Bromophenyl)oxetane-3-carboxylic acid ≥95%
3-(3-Bromophenyl)oxetane-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₉BrO₃
MDL Number: MFCD22546748
CAS Number: 1363381-80-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR