3-(4-Bromophenyl)oxetane ≥95%

Supplier: Matrix Scientific

115856-250MG 115856-500MG 115856-1G
102898-568EA 1718.46 USD
102898-568 102898-570 102898-572
3-(4-Bromophenyl)oxetane ≥95%
3-(4-Bromophenyl)oxetane

Hazard: Irritant

Formula: C₉H₉BrO
MDL Number: MFCD22628728
CAS Number: 1402158-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR