3-(3-Bromophenyl)oxetane ≥95%

Supplier: Matrix Scientific

115855-250MG 115855-500MG 115855-1G
102898-562EA 1098.46 USD
102898-562 102898-564 102898-566
3-(3-Bromophenyl)oxetane ≥95%
3-(3-Bromophenyl)oxetane

Hazard: Irritant

Formula: C₉H₉BrO
MDL Number: MFCD17012864
CAS Number: 1044507-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR