3-(3-Bromophenyl)-3-(hydroxymethyl)oxetane ≥95%

Supplier: Matrix Scientific

115853-250MG 115853-500MG 115853-1G
102898-550EA 1593.85 USD
102898-550 102898-552 102898-554
3-(3-Bromophenyl)-3-(hydroxymethyl)oxetane ≥95%
3-(3-Bromophenyl)-3-(hydroxymethyl)oxetane

Hazard: Irritant

Formula: C₁₀H₁₁BrO₂
MDL Number: MFCD22566188
CAS Number: 1363382-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR