N-(3-Bromobenzyl)oxetan-3-amine ≥95%

Supplier: Matrix Scientific

115848-1G 115848-5G 115848-10G
102898-474EA 1373.85 USD
102898-474 102898-476 102898-478
N-(3-Bromobenzyl)oxetan-3-amine ≥95%
N-(3-Bromobenzyl)oxetan-3-amine

Hazard: Irritant

Formula: C₁₀H₁₂BrNO
MDL Number: MFCD18849410
CAS Number: 1339688-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR