N-(2-Bromobenzyl)oxetan-3-amine ≥95%

Supplier: Matrix Scientific

115847-1G 115847-5G 115847-10G
102898-468EA 1373.85 USD
102898-468 102898-470 102898-472
N-(2-Bromobenzyl)oxetan-3-amine ≥95%
N-(2-Bromobenzyl)oxetan-3-amine

Hazard: Irritant

Formula: C₁₀H₁₂BrNO
MDL Number: MFCD18849411
CAS Number: 1339664-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR