7-Boc-1-oxa-3-oxo-7-azaspiro[3.5]nonane ≥95%

Supplier: Matrix Scientific

115846-250MG 115846-500MG 115846-1G
102898-522EA 2061.54 USD
102898-522 102898-524 102898-526
7-Boc-1-oxa-3-oxo-7-azaspiro[3.5]nonane ≥95%
7-Boc-1-oxa-3-oxo-7-azaspiro[3.5]nonane

Hazard: Irritant

Formula: C₁₂H₁₉NO₄
MW: 241.29 g/mol
MDL Number: MFCD22628723
CAS Number: 1408076-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR