8-Boc-2-oxa-5,8-diazaspiro[3.5]nonane ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 2-oxa-5,8-diazaspiro[3.5]nonane-8-carboxylate

115845-250MG 115845-500MG 115845-1G
102898-516EA 1992.31 USD
102898-516 102898-518 102898-520
8-Boc-2-oxa-5,8-diazaspiro[3.5]nonane ≥95%
8-Boc-2-oxa-5,8-diazaspiro[3.5]nonane

Hazard: Irritant

Formula: C₁₁H₂₀N₂O₃
MW: 228.29 g/mol
MDL Number: MFCD22035557
CAS Number: 1367777-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR