3-Boc-amino-3-(4-cyanophenyl)oxetane ≥95%

Supplier: Matrix Scientific

115840-250MG 115840-500MG
102898-324EA 2196.92 USD
102898-324 102900-300
3-Boc-amino-3-(4-cyanophenyl)oxetane ≥95%
3-Boc-amino-3-(4-cyanophenyl)oxetane

Hazard: Irritant

Formula: C₁₅H₁₈N₂O₃
MDL Number: MFCD17215904
CAS Number: 1158098-77-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR