(S)-Benzyl-1-(oxetan-3-yl)pyrrolidin-3-ylcarbamate ≥95%
Supplier: Matrix Scientific
115836-500MG
115836-1G
115836-5G
102898-300EA
1238.46
USD
102898-300
102898-302
102898-304
(S)-Benzyl-1-(oxetan-3-yl)pyrrolidin-3-ylcarbamate ≥95%
(S)-Benzyl-1-(oxetan-3-yl)pyrrolidin-3-ylcarbamate
Hazard: Irritant
Formula:
C₁₅H₂₀N₂O₃ |
MDL Number:
MFCD21602105 CAS Number: 1349807-52-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...