(R)-Benzyl-1-(oxetan-3-yl)piperidin-3-ylcarbamate ≥95%

Supplier: Matrix Scientific

115834-5G 115834-10G
102898-110EA 6152.31 USD
102898-110 102898-112
(R)-Benzyl-1-(oxetan-3-yl)piperidin-3-ylcarbamate ≥95%
(R)-Benzyl-1-(oxetan-3-yl)piperidin-3-ylcarbamate

Hazard: Irritant

Formula: C₁₆H₂₂N₂O₃
MDL Number: MFCD20233247
CAS Number: 1349699-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR