Benzyl-N-(3-ethyloxetan-3-yl)carbamate ≥95%

Supplier: Matrix Scientific

115831-1G 115831-10G
102900-280EA 1787.69 USD
102900-280 102900-284
Benzyl-N-(3-ethyloxetan-3-yl)carbamate ≥95%
Benzyl-N-(3-ethyloxetan-3-yl)carbamate

Hazard: Irritant

Formula: C₁₃H₁₇NO₃
MW: 235.29 g/mol
MDL Number: MFCD21602199
CAS Number: 1365969-56-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR