3-Aminomethyl-3-fluorooxetane ≥90%

Supplier: Matrix Scientific

Synonyms: (3-Fluorooxetan-3-yl)methanamine

115829-250MG 115829-1G
102900-268EA 1663.08 USD
102900-268 102900-272
3-Aminomethyl-3-fluorooxetane ≥90%
3-Aminomethyl-3-fluorooxetane

Hazard: Irritant

Formula: C₄H₈FNO
MW: 105.11 g/mol
MDL Number: MFCD18073259
CAS Number: 883311-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR