3-Amino-3-(3-bromophenyl)oxetane hydrochloride ≥95%

Supplier: Matrix Scientific

115827-250MG 115827-500MG 115827-1G
102898-282EA 2267.69 USD
102898-282 102899-754 102899-756
3-Amino-3-(3-bromophenyl)oxetane hydrochloride ≥95%
3-Amino-3-(3-bromophenyl)oxetane hydrochloride

Hazard: Irritant

Formula: C₉H₁₁BrClNO
MDL Number: MFCD19381731
CAS Number: 1332920-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR