3-Cyano-3-(pyrrolidin-1-yl)oxetane ≥95%

Supplier: Matrix Scientific

115816-1G 115816-5G 115816-10G
102898-222EA 1373.85 USD
102898-222 102899-700 102899-702
3-Cyano-3-(pyrrolidin-1-yl)oxetane ≥95%
3-Cyano-3-(pyrrolidin-1-yl)oxetane

Hazard: Irritant

Formula: C₈H₁₂N₂O
MDL Number: MFCD22575229
CAS Number: 1208984-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR