(3,5-Dichloro-4-pyridyl)methanol ≥98%

Supplier: Matrix Scientific

Synonyms: (3,5-Dichloropyridin-4-yl)methanol

115812-500MG 115812-1G 115812-5G
102898-198EA 413.85 USD
102898-198 102898-200 102898-202
(3,5-Dichloro-4-pyridyl)methanol ≥98%
(3,5-Dichloro-4-pyridyl)methanol

Hazard: Irritant

Formula: C₆H₅Cl₂NO
MW: 178.02 g/mol
MDL Number: MFCD06202840
CAS Number: 159783-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR