1-(2-Chloroethyl)-4-ethyl-1,4-dihydro-5H-tetrazol-5-one ≥95%

Supplier: Matrix Scientific

115811-1G 115811-5G 115811-10G
102898-192EA 604.62 USD
102898-192 102898-194 102898-196
1-(2-Chloroethyl)-4-ethyl-1,4-dihydro-5H-tetrazol-5-one ≥95%
1-(2-Chloroethyl)-4-ethyl-1,4-dihydro-5H-tetrazol-5-one

Hazard: Irritant

Formula: C₅H₉ClN₄O
MDL Number: MFCD09907794
CAS Number: 69049-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR