3-Bromothieno[3,2-c]pyridin-4-amine ≥95%

Supplier: Matrix Scientific

115806-500MG 115806-1G
102898-170EA 1044.62 USD
102898-170 102898-172
3-Bromothieno[3,2-c]pyridin-4-amine ≥95%
3-Bromothieno[3,2-c]pyridin-4-amine

Hazard: Irritant

Formula: C7H5BrN2S
MW: 229.0970 g/mol
MDL Number: MFCD08275115
CAS Number: 799293-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR