(S)-Piperidine-3-carboxamide hydrochloride ≥95%

Supplier: Matrix Scientific

115800-500MG 115800-1G
102898-140EA 769.23 USD
102898-140 102898-142
(S)-Piperidine-3-carboxamide hydrochloride ≥95%
(S)-Piperidine-3-carboxamide hydrochloride

Hazard: Irritant

Formula: C₆H₁₃ClN₂O
MDL Number: MFCD22415259
CAS Number: 1026661-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR