(S)-4-Oxo-piperidine-2-carboxylic acid methyl ester ≥95%

Supplier: Matrix Scientific

115798-250MG 115798-500MG 115798-1G
102899-694EA 1593.85 USD
102899-694 102899-696 102899-698
(S)-4-Oxo-piperidine-2-carboxylic acid methyl ester ≥95%
(S)-4-Oxo-piperidine-2-carboxylic acid methyl ester

Hazard: Irritant

Formula: C₇H₁₁NO₃
MDL Number: MFCD13175034
CAS Number: 761360-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR