6-Oxa-1-azaspiro[3.4]octane oxalate ≥95%

Supplier: Matrix Scientific

115796-250MG 115796-500MG 115796-1G
102899-684EA 1443.08 USD
102899-684 102899-686 102899-688
6-Oxa-1-azaspiro[3.4]octane oxalate ≥95%
6-Oxa-1-azaspiro[3.4]octane oxalate

Hazard: Irritant

Formula: C₆H₁₁NO
MDL Number: MFCD22415291
CAS Number: 71850-23-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR