3-(m-Tolyl)oxetan-3-amine hydrochloride ≥95%

Supplier: Matrix Scientific

115792-1G 115792-5G 115792-10G
102897-646EA 1787.69 USD
102897-646 102897-648 102897-650
3-(m-Tolyl)oxetan-3-amine hydrochloride ≥95%
3-(m-Tolyl)oxetan-3-amine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₄ClNO
MDL Number: MFCD19381736
CAS Number: 1322200-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR