(3-tert-Butoxycarbonylaminooxetan-3-yl)acetic acid ≥95%

Supplier: Matrix Scientific

115790-250MG 115790-500MG 115790-1G
102897-568EA 893.85 USD
102897-568 102899-670 102899-672
(3-tert-Butoxycarbonylaminooxetan-3-yl)acetic acid ≥95%
(3-tert-Butoxycarbonylaminooxetan-3-yl)acetic acid

Hazard: Irritant

Formula: C₁₀H₁₇NO₅
MW: 231.25 g/mol
MDL Number: MFCD16140246
CAS Number: 1221715-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR