3-(Oxetan-3-yl)pyrrolidine ≥95%

Supplier: Matrix Scientific

115789-250MG 115789-500MG 115789-1G
102897-562EA 1690.77 USD
102897-562 102897-564 102897-566
3-(Oxetan-3-yl)pyrrolidine ≥95%
3-(Oxetan-3-yl)pyrrolidine

Hazard: Irritant

Formula: C₇H₁₃NO
MDL Number: MFCD18793263
CAS Number: 1273567-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR