(3S)-1-(Oxetan-3-yl)pyrrolidin-3-amine ≥95%
Supplier: Matrix Scientific
115788-1G
115788-5G
115788-10G
102897-556EA
2363.08
USD
102897-556
102897-558
102897-560
(3S)-1-(Oxetan-3-yl)pyrrolidin-3-amine ≥95%
(3S)-1-(Oxetan-3-yl)pyrrolidin-3-amine
Hazard: Irritant
Formula:
C₇H₁₄N₂O |
MDL Number:
MFCD19690700 CAS Number: 1256667-60-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...