(3R)-1-(Oxetan-3-yl)pyrrolidin-3-amine ≥95%
Supplier: Matrix Scientific
115787-1G
115787-5G
115787-10G
102897-550EA
2363.08
USD
102897-550
102897-552
102897-554
(3R)-1-(Oxetan-3-yl)pyrrolidin-3-amine ≥95%
(3R)-1-(Oxetan-3-yl)pyrrolidin-3-amine
Hazard: Irritant
Formula:
C₇H₁₄N₂O |
MDL Number:
MFCD19690701 CAS Number: 1256667-56-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...