2-Oxetan-3-yl-2H-pyrazol-3-ylamine ≥95%

Supplier: Matrix Scientific

115786-250MG 115786-500MG 115786-1G
102897-544EA 909.23 USD
102897-544 102897-546 102897-548
2-Oxetan-3-yl-2H-pyrazol-3-ylamine ≥95%
2-Oxetan-3-yl-2H-pyrazol-3-ylamine

Hazard: Irritant

Formula: C₆H₉N₃O
MDL Number: MFCD20233066
CAS Number: 1349708-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR