(2-Hydroxy-4-methylphenyl)boronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Hydroxy-4-methylphenyl boronic acid

115780-250MG 115780-500MG 115780-1G
102897-512EA 1458.46 USD
102897-512 102897-514 102897-516
(2-Hydroxy-4-methylphenyl)boronic acid ≥97%
(2-Hydroxy-4-methylphenyl)boronic acid

Hazard: Irritant

Formula: C₇H₉BO₃
MW: 151.96 g/mol
MDL Number: MFCD18397304
CAS Number: 259209-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR