Ethyl-7-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate ≥95%
Supplier: Matrix Scientific
115776-250MG
115776-500MG
115776-1G
102897-492EA
1443.08
USD
102897-492
102897-494
102897-496
Ethyl-7-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate ≥95%
Ethyl-7-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate
Hazard: Irritant
Formula:
C₈H₆BrN₃O₂ |
CAS Number:
1363382-48-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...