Diphenyl[(E/Z)-3,3,3-trifluoroprop-1-enyl]sulfonium trifluoromethanesulfonate ≥95%

Supplier: Matrix Scientific

115774-500MG 115774-1G 115774-5G
102899-664EA 769.23 USD
102899-664 102899-666 102899-668
Diphenyl[(E/Z)-3,3,3-trifluoroprop-1-enyl]sulfonium trifluoromethanesulfonate ≥95%
Diphenyl[(E/Z)-3,3,3-trifluoroprop-1-enyl]sulfonium trifluoromethanesulfonate

Hazard: Irritant

Formula: C₁₆H₁₂F₆O₃S₂
MDL Number: MFCD22573385
CAS Number: 1226574-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR