Diethyl-9-azabicyclo[3.3.1]nonane-3,7-dicarboxylate hydrochloride ≥95%
Supplier: Matrix Scientific
115768-250MG
115768-500MG
115768-1G
102897-320EA
2541.54
USD
102897-320
102897-472
102897-474
Diethyl-9-azabicyclo[3.3.1]nonane-3,7-dicarboxylate hydrochloride ≥95%
Diethyl-9-azabicyclo[3.3.1]nonane-3,7-dicarboxylate hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₄ClNO₄ |
MDL Number:
MFCD22573379 CAS Number: 1363380-60-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...