2-Cyano-4,5-dimethylthiazole ≥97%

Supplier: Matrix Scientific

115767-500MG 115767-1G 115767-250MG
102897-316EA 2455.38 USD
102897-316 102897-318 102897-470
2-Cyano-4,5-dimethylthiazole ≥97%
2-Cyano-4,5-dimethylthiazole

Hazard: Irritant

Formula: C₆H₆N₂S
MDL Number: MFCD19440547
CAS Number: 1378802-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR