exo-3-Cyano-9-azabicyclo-[3.3.1]nonane hydrochloride ≥95%
Supplier: Matrix Scientific
115766-250MG
115766-500MG
115766-1G
102897-464EA
963.08
USD
102897-464
102897-466
102897-468
exo-3-Cyano-9-azabicyclo-[3.3.1]nonane hydrochloride ≥95%
exo-3-Cyano-9-azabicyclo-[3.3.1]nonane hydrochloride
Hazard: Irritant
Formula:
C₉H₁₄N₂ |
MDL Number:
MFCD22415209 CAS Number: 1363383-01-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...