4-Chloro-5-(methylsulfonyl)-7H-pyrrolo[2,3-d]pyrimidine ≥95%
Supplier: Matrix Scientific
115763-250MG
115763-500MG
115763-1G
102897-448EA
1867.69
USD
102897-448
102897-450
102897-452
4-Chloro-5-(methylsulfonyl)-7H-pyrrolo[2,3-d]pyrimidine ≥95%
4-Chloro-5-(methylsulfonyl)-7H-pyrrolo[2,3-d]pyrimidine
Hazard: Irritant
Formula:
C₇H₆ClN₃O₂S |
MDL Number:
MFCD22415205 CAS Number: 1363381-40-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...