3-(5-Bromopyridin-3-yl)azetidine-1-carboxylic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

115761-500MG 115761-1G
102897-438EA 1318.46 USD
102897-438 102897-440
3-(5-Bromopyridin-3-yl)azetidine-1-carboxylic acid tert-butyl ester ≥95%
3-(5-Bromopyridin-3-yl)azetidine-1-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₃H₁₇BrN₂O₂
MDL Number: MFCD22573374
CAS Number: 1392803-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR