5-Bromo-3-nitro-1H-pyrrolo[2,3-b]pyridine ≥95%

Supplier: Matrix Scientific

115760-1G 115760-5G 115760-25G
102897-432EA 413.85 USD
102897-432 102897-434 102897-436
5-Bromo-3-nitro-1H-pyrrolo[2,3-b]pyridine ≥95%
5-Bromo-3-nitro-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₇H₄BrN₃O₂
MDL Number: MFCD12405111
CAS Number: 507462-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR