(2S)-1-Boc-aziridine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

115757-250MG 115757-500MG 115757-1G
102897-414EA 1318.46 USD
102897-414 102897-416 102897-418
(2S)-1-Boc-aziridine-2-carboxylic acid ≥95%
(2S)-1-Boc-aziridine-2-carboxylic acid

Hazard: Irritant

Formula: C₈H₁₃NO₄
MDL Number: MFCD11519231
CAS Number: 1315000-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR