3-Boc-3-azabicyclo[3.1.1]heptane-6-carboxamide ≥95%

Supplier: Matrix Scientific

115751-500MG 115751-1G
102897-380EA 1867.69 USD
102897-380 102897-382
3-Boc-3-azabicyclo[3.1.1]heptane-6-carboxamide ≥95%
3-Boc-3-azabicyclo[3.1.1]heptane-6-carboxamide

Hazard: Irritant

Formula: C₁₂H₂₀N₂O₃
CAS Number: 1392803-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR