5-Aminothiazole-2-carboxylic acid hydrochloride ≥97%

Supplier: Matrix Scientific

115749-250MG 115749-500MG 115749-1G
102897-370EA 1318.46 USD
102897-370 102897-372 102897-374
5-Aminothiazole-2-carboxylic acid hydrochloride ≥97%
5-Aminothiazole-2-carboxylic acid hydrochloride

Hazard: Irritant

Formula: C₄H₅ClN₂O₂S
MDL Number: MFCD22573368
CAS Number: 1404373-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR