3-Aminomethyl-5-(methoxymethyl)pyrazole hydrochloride ≥95%
Supplier: Matrix Scientific
115747-250MG
115747-500MG
115747-1G
102897-360EA
1593.85
USD
102897-360
102897-362
102897-364
3-Aminomethyl-5-(methoxymethyl)pyrazole hydrochloride ≥95%
3-Aminomethyl-5-(methoxymethyl)pyrazole hydrochloride
Hazard: Irritant
Formula:
C₅H₁₀ClN₃O |
CAS Number:
1297607-82-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...