3-Amino-5-methoxypyridine hydrochloride ≥95%

Supplier: Matrix Scientific

115745-500MG 115745-1G
102897-350EA 344.62 USD
102897-350 102897-352
3-Amino-5-methoxypyridine hydrochloride ≥95%
3-Amino-5-methoxypyridine hydrochloride

Hazard: Irritant

Formula: C₆H₉ClN₂O
MDL Number: MFCD12913792
CAS Number: 896160-77-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR